Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- WRKY59
- Entrez Gene ID
-
816726
- Gene Name
- WRKY DNA-binding protein 59
- Gene Aliases
-
AT2G21900, ATWRKY59, F7D8.22, F7D8_22, WRKY DNA-binding protein 59
- Chromosome Location
-
Chr.2: 9333844 - 9336278 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_179688.2 |
NP_850019.1 |
2 - 3 |
443 |
62 |
View Full Product Details