Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PFK5
Entrez Gene ID
816781
Gene Name
phosphofructokinase 5
Gene Aliases
AT2G22480, F14M13.12, F14M13_12, phosphofructokinase 5
Chromosome Location
Chr.2: 9545283 - 9548640 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_179694.4 NP_850025.1 6 - 7 1184 65
GenBank mRNA AY070402.1 - 6 - 7 852 65
AY096578.1 - 6 - 7 797 65

View Full Product Details