Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RTFL2
Entrez Gene ID
817461
Gene Name
ROTUNDIFOLIA like 2
Gene Aliases
AT2G29125, DEVIL 13, DVL13, ROTUNDIFOLIA like 2
Chromosome Location
Chr.2: 12523276 - 12524193 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_179799.2 NP_850130.1 1 - 1 296 104
GenBank mRNA AY136336.1 - 1 - 1 230 104
BT000101.1 - 1 - 1 100 104

View Full Product Details