Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBP47B
Entrez Gene ID
821447
Gene Name
RNA-binding protein 47B
Gene Aliases
AT3G19130, ATRBP47B, RNA-binding protein 47B
Chromosome Location
Chr.3: 6611182 - 6614107 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112800.4 NP_188544.1 3 - 4 1247 120
GenBank mRNA AK226472.1 - 3 - 4 1188 120
BT026489.1 - 3 - 4 963 120

View Full Product Details