Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAD54
Entrez Gene ID
821455
Gene Name
DNA repair/recombination protein
Gene Aliases
AT3G19210, ATRAD54, CHR25, homolog of RAD54
Chromosome Location
Chr.3: 6652695 - 6658988 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203004.1 NP_001189933.1 12 - 13 1663 96
NM_001338387.1 NP_001325757.1 12 - 13 1644 96
NM_112808.4 NP_188552.3 12 - 13 1644 96
GenBank mRNA AB250666.1 - 12 - 13 1619 96
DQ912973.1 - 12 - 13 1557 96

View Full Product Details