Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAP18
Entrez Gene ID
821596
Gene Name
purple acid phosphatase 18
Gene Aliases
AT3G20500, ATPAP18, PURPLE ACID PHOSPHATASE 18, purple acid phosphatase 18
Chromosome Location
Chr.3: 7157900 - 7160446 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112942.5 NP_188686.2 2 - 3 839 89
GenBank mRNA AY062488.1 - 2 - 3 828 89
AY093272.1 - 2 - 3 813 89

View Full Product Details