Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DDL
Entrez Gene ID
821601
Gene Name
SMAD/FHA domain-containing protein
Gene Aliases
AT3G20550, DAWDLE
Chromosome Location
Chr.3: 7174436 - 7178020 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112947.4 NP_188691.1 4 - 5 653 80
GenBank mRNA AY062626.1 - 4 - 5 570 80
BT000005.1 - 3 - 4 470 80

View Full Product Details