Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DRL1
Entrez Gene ID
829695
Gene Name
dihydroflavonol 4-reductase-like1
Gene Aliases
AT4G35420, F15J1.1, TKPR1, dihydroflavonol 4-reductase-like1, tetraketide alpha-pyrone reductase 1
Chromosome Location
Chr.4: 16833933 - 16835619 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119708.3 NP_195268.2 3 - 4 481 76
GenBank mRNA BT022119.1 - 3 - 4 477 76
BT025661.1 - 3 - 4 473 76

View Full Product Details