Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCB1
Entrez Gene ID
829800
Gene Name
long-chain base1
Gene Aliases
AP22.60, AP22_60, AT4G36480, ATLCB1, EMB2779, EMBRYO DEFECTIVE 2779, FBR11, FUMONISIN B1 RESISTANT 11, long-chain base1
Chromosome Location
Chr.4: 17217909 - 17221625 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036719.2 NP_001031796.1 6 - 7 670 127
NM_001342410.1 NP_001329288.1 6 - 7 940 127
NM_119811.3 NP_568005.1 6 - 7 940 127
GenBank mRNA AB063254.1 - 5 - 6 482 127
AY120759.1 - 6 - 7 758 127
BT000131.1 - 5 - 6 461 127

View Full Product Details