Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PBRP
Entrez Gene ID
829817
Gene Name
plant-specific TFIIB-related protein
Gene Aliases
AP22.23, AP22_23, AT4G36650, ATPBRP, plant-specific TFIIB-related protein
Chromosome Location
Chr.4: 17283033 - 17285656 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085033.4 NP_001078502.4 1 - 2 607 70
NM_119828.6 NP_195383.1 1 - 2 713 70
GenBank mRNA AJ295068.1 - 1 - 2 656 70
AY463603.1 - 1 - 2 584 70
BT006470.1 - 1 - 2 584 70

View Full Product Details