Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BT5
Entrez Gene ID
829915
Gene Name
BTB and TAZ domain protein 5
Gene Aliases
AT4G37610, BTB and TAZ domain protein 5, F19F18.100, F19F18_100
Chromosome Location
Chr.4: 17670481 - 17672353 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119924.3 NP_568031.1 3 - 4 562 112
GenBank mRNA AY316678.1 - 2 - 3 356 112
BT025247.1 - 2 - 3 356 112

View Full Product Details