Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRF2
Entrez Gene ID
829930
Gene Name
growth-regulating factor 2
Gene Aliases
AT4G37740, AtGRF2, T28I19.20, T28I19_20, growth-regulating factor 2
Chromosome Location
Chr.4: 17725337 - 17727909 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119936.5 NP_195488.2 2 - 3 937 104
GenBank mRNA AB493726.1 - 2 - 3 637 104
AK176372.1 - 2 - 3 758 104
AK176432.1 - 2 - 3 687 104
AY102635.1 - 2 - 3 711 104

View Full Product Details