Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANT
Entrez Gene ID
829931
Gene Name
Integrase-type DNA-binding superfamily protein
Gene Aliases
AINTEGUMENTA, AT4G37750, AtANT, CKC, CKC1, COMPLEMENTING A PROTEIN KINASE C MUTANT 1, DRAGON, DRG, T28I19.30, T28I19_30
Chromosome Location
Chr.4: 17739514 - 17742407 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119937.3 NP_195489.1 2 - 3 1125 116
GenBank mRNA AY080706.1 - 2 - 3 1014 116
AY117207.1 - 2 - 3 857 116
U40256.1 - 2 - 3 1125 116
U41339.1 - 2 - 3 894 116

View Full Product Details