Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP78A8
Entrez Gene ID
839233
Gene Name
cytochrome P450, family 78, subfamily A, polypeptide 8
Gene Aliases
''cytochrome P450, AT1G01190, F6F3.25, cytochrome P450, family 78, polypeptide 8, polypeptide 8'', subfamily A
Chromosome Location
Chr.1: 82984 - 84946 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331253.1 NP_001318901.1 1 - 2 977 114
NM_100001.2 NP_171627.2 1 - 2 1059 114

View Full Product Details