Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- FRO1
- Entrez Gene ID
-
837194
- Gene Name
- ferric reduction oxidase 1
- Gene Aliases
-
AT1G01590, ATFRO1, F22L4.13, F22L4_13, FERRIC REDUCTION OXIDASE 1, ferric reduction oxidase 1
- Chromosome Location
-
Chr.1: 214150 - 217734 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_100041.2 |
NP_171665.1 |
2 - 3 |
356 |
94 |
View Full Product Details