Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RALF1
- Entrez Gene ID
-
839389
- Gene Name
- rapid alkalinization factor 1
- Gene Aliases
-
AT1G02900, ATRALF1, F22D16.10, F22D16_10, RALF-LIKE 1, RALFL1, RAPID ALKALINIZATION FACTOR 1, rapid alkalinization factor 1
- Chromosome Location
-
Chr.1: 653604 - 654457 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_100171.3 |
NP_171789.1 |
1 - 1 |
543 |
131 |
View Full Product Details