Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GSTF5
Entrez Gene ID
839479
Gene Name
glutathione S-transferase (class phi) 5
Gene Aliases
AT1G02940, ATGSTF5, F22D16.6, F22D16_6, GLUTATHIONE S-TRANSFERASE (CLASS PHI) 5, glutathione S-transferase (class phi) 5
Chromosome Location
Chr.1: 662963 - 664290 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331365.1 NP_001322019.1 2 - 3 199 78
NM_001331366.1 NP_001322018.1 2 - 3 170 78
NM_001331367.1 NP_001322017.1 2 - 3 131 78
NM_100175.2 NP_171793.1 1 - 2 111 78

View Full Product Details