Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TPS7
Entrez Gene ID
837152
Gene Name
trehalose-phosphatase/synthase 7
Gene Aliases
AT1G06410, ATTPS7, ATTPSA, T2D23.11, T2D23_11, TREHALOSE -6-PHOSPHATASE SYNTHASE S7, trehalose-phosphatase/synthase 7
Chromosome Location
Chr.1: 1954674 - 1958866 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331626.1 NP_001321476.1 3 - 4 2519 71
NM_001331627.1 NP_001321475.1 3 - 4 2598 71
NM_100521.3 NP_172129.1 3 - 4 2521 71

View Full Product Details