Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GAUT6
Entrez Gene ID
837189
Gene Name
galacturonosyltransferase 6
Gene Aliases
AT1G06780, F4H5.13, F4H5_13, galacturonosyltransferase 6
Chromosome Location
Chr.1: 2083311 - 2087141 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100555.3 NP_563771.1 2 - 3 483 90
GenBank mRNA AY045900.1 - 2 - 3 415 90
AY091452.1 - 2 - 3 150 90
KJ139026.1 - 2 - 3 150 90

View Full Product Details