Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARAC9
Entrez Gene ID
819077
Gene Name
RAC-like 9
Gene Aliases
AT2G44690, ATROP8, Arabidopsis RAC-like 9, F16B22.18, RHO-RELATED PROTEIN FROM PLANTS 8, ROP8
Chromosome Location
Chr.2: 18429121 - 18430882 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130033.3 NP_566024.1 4 - 5 546 67
GenBank mRNA AF156896.1 - 4 - 5 403 67
BT025645.1 - 4 - 5 391 67

View Full Product Details