Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AUR3
Entrez Gene ID
819157
Gene Name
ataurora3
Gene Aliases
AT2G45490, AtAUR3, F17K2.2, ataurora3
Chromosome Location
Chr.2: 18747509 - 18749215 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337135.1 NP_001323763.1 1 - 2 257 68
NM_130111.4 NP_182073.1 1 - 2 294 68
GenBank mRNA AJ854185.1 - 1 - 2 123 68
AY074653.1 - 1 - 2 154 68

View Full Product Details