Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AUR3
Entrez Gene ID
819157
Gene Name
ataurora3
Gene Aliases
AT2G45490, AtAUR3, F17K2.2, ataurora3
Chromosome Location
Chr.2: 18747509 - 18749215 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337135.1 NP_001323763.1 4 - 5 807 75
NM_130111.4 NP_182073.1 4 - 5 844 75
GenBank mRNA AJ854185.1 - 4 - 5 673 75
AY074653.1 - 4 - 5 704 75

View Full Product Details