The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130140.3 | NP_566056.1 | 5 - 6 | 746 | 81 |
GenBank mRNA | AF360125.1 | - | 5 - 6 | 616 | 81 |
AJ010820.1 | - | 5 - 6 | 596 | 81 | |
AY051026.1 | - | 5 - 6 | 586 | 81 |