Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPX4
Entrez Gene ID
819427
Gene Name
glutathione peroxidase 4
Gene Aliases
AT2G48150, ATGPX4, F11L15.5, glutathione peroxidase 4
Chromosome Location
Chr.2: 19687831 - 19689229 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337302.1 NP_001324284.1 1 - 2 363 67
NM_130382.2 NP_566128.1 2 - 3 263 67
GenBank mRNA AK118448.1 - 2 - 3 184 67
AY088692.1 - 2 - 3 207 67
BT003735.1 - 2 - 3 133 67

View Full Product Details