Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FATB
Entrez Gene ID
837372
Gene Name
fatty acyl-ACP thioesterases B
Gene Aliases
AT1G08510, T27G7.19, T27G7_19, fatty acyl-ACP thioesterases B
Chromosome Location
Chr.1: 2691062 - 2694547 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100724.3 NP_172327.1 5 - 6 1433 101
GenBank mRNA AY054679.1 - 5 - 6 1256 101
AY059827.1 - 5 - 6 1057 101
AY064652.1 - 4 - 5 915 101
BT008505.1 - 4 - 5 915 101

View Full Product Details