Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EDR1
Entrez Gene ID
837393
Gene Name
Protein kinase superfamily protein
Gene Aliases
AT1G08720, ATEDR1, ENHANCED DISEASE RESISTANCE 1, F22O13.20, F22O13_20, MAPKK KINASE EDR1
Chromosome Location
Chr.1: 2773941 - 2779308 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331786.1 NP_001322729.1 3 - 4 1086 75
NM_100745.2 NP_563824.1 3 - 4 1086 75
GenBank mRNA AF305913.1 - 3 - 4 997 75

View Full Product Details