Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VAP27-2
Entrez Gene ID
837404
Gene Name
vamp/synaptobrevin-associated protein 27-2
Gene Aliases
AT1G08820, F22O13.31, F22O13_31, vamp/synaptobrevin-associated protein 27-2
Chromosome Location
Chr.1: 2821354 - 2825481 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035927.2 NP_001031004.1 3 - 4 496 88
NM_001331791.1 NP_001321657.1 3 - 4 1089 88
NM_001331792.1 NP_001321655.1 3 - 4 935 88
NM_001331793.1 NP_001321656.1 3 - 4 871 88
NM_100756.2 NP_172359.2 4 - 5 414 88
GenBank mRNA AK317489.1 - 3 - 4 269 88
AK317626.1 - 4 - 5 415 88

View Full Product Details