Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VAP27-2
Entrez Gene ID
837404
Gene Name
vamp/synaptobrevin-associated protein 27-2
Gene Aliases
AT1G08820, F22O13.31, F22O13_31, vamp/synaptobrevin-associated protein 27-2
Chromosome Location
Chr.1: 2821354 - 2825481 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035927.2 NP_001031004.1 4 - 5 580 152
NM_001331791.1 NP_001321657.1 4 - 5 1173 152
NM_001331792.1 NP_001321655.1 4 - 5 1019 152
NM_001331793.1 NP_001321656.1 4 - 5 955 152
NM_100756.2 NP_172359.2 5 - 6 498 152
GenBank mRNA AK317489.1 - 4 - 5 353 152
AK317626.1 - 5 - 6 499 152

View Full Product Details