Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BIP3
Entrez Gene ID
837429
Gene Name
Heat shock protein 70 (Hsp 70) family protein
Gene Aliases
AT1G09080, F7G19.5, F7G19_5, binding protein 3
Chromosome Location
Chr.1: 2929217 - 2931850 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198015.2 NP_001184944.1 4 - 5 1320 92
NM_100779.5 NP_172382.4 4 - 5 1341 92
GenBank mRNA AY156728.1 - 4 - 5 1341 92

View Full Product Details