Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PUB18
Entrez Gene ID
837597
Gene Name
plant U-box 18
Gene Aliases
ARABIDOPSIS THALIANA PLANT U-BOX 18, AT1G10560, ATPUB18, T10O24.19, T10O24_19, plant U-box 18
Chromosome Location
Chr.1: 3484399 - 3486965 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100931.3 NP_172526.1 1 - 1 2435 178

View Full Product Details