Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BGAL4
Entrez Gene ID
835789
Gene Name
beta-galactosidase 4
Gene Aliases
AT5G56870, MPI10.3, MPI10_3, beta-galactosidase 4
Chromosome Location
Chr.5: 23003909 - 23009672 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125070.3 NP_200498.1 13 - 14 1912 63
GenBank mRNA AJ270300.1 - 13 - 14 1568 63

View Full Product Details