Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NAP1;3
Entrez Gene ID
835797
Gene Name
nucleosome assembly protein 1;3
Gene Aliases
AT5G56950, MHM17.6, MHM17_6, NFA03, NFA3, NUCLEOSOME/CHROMATIN ASSEMBLY FACTOR GROUP A 03, NUCLEOSOME/CHROMATIN ASSEMBLY FACTOR GROUP A3, nucleosome assembly protein 1;3
Chromosome Location
Chr.5: 23032449 - 23035693 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125077.4 NP_568844.1 10 - 11 1207 147
GenBank mRNA AF370487.1 - 10 - 11 1175 147
AY064641.1 - 10 - 11 1038 147

View Full Product Details