Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HDA05
Entrez Gene ID
836227
Gene Name
histone deacetylase 5
Gene Aliases
AT5G61060, ATHDA5, HDA5, MAF19.7, MAF19_7, histone deacetylase 5
Chromosome Location
Chr.5: 24566783 - 24571072 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203654.1 NP_001190583.1 4 - 5 734 97
NM_125499.5 NP_200914.2 4 - 5 815 97
GenBank mRNA AY090936.1 - 4 - 5 734 97
AY120784.1 - 4 - 5 728 97
BT000073.1 - 4 - 5 660 97

View Full Product Details