Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RR23
Entrez Gene ID
836332
Gene Name
response regulator 23
Gene Aliases
ARR23, AT5G62120, MTG10.15, MTG10_15, response regulator 23
Chromosome Location
Chr.5: 24946955 - 24948225 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125605.1 NP_201018.1 2 - 3 279 85

View Full Product Details