Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ERL1
Entrez Gene ID
836344
Gene Name
ERECTA-like 1
Gene Aliases
AT5G62230, ERECTA-like 1, MMI9.14, MMI9_14
Chromosome Location
Chr.5: 24996129 - 25002439 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203666.1 NP_001190595.1 5 - 6 740 138
NM_125617.3 NP_201029.1 5 - 6 733 138
GenBank mRNA FJ708810.1 - 5 - 6 436 138

View Full Product Details