Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBP47A
Entrez Gene ID
841384
Gene Name
RNA-binding protein 47A
Gene Aliases
AT1G49600, ATRBP47A, F14J22.16, F14J22_16, RNA-binding protein 47A
Chromosome Location
Chr.1: 18356799 - 18360447 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333392.1 NP_001321910.1 4 - 5 1393 91
NM_001333393.1 NP_001321909.1 4 - 5 1414 91
NM_103848.3 NP_175383.1 4 - 5 1414 91

View Full Product Details