Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPK33
Entrez Gene ID
841492
Gene Name
calcium-dependent protein kinase 33
Gene Aliases
AT1G50700, F17J6.22, F17J6_22, calcium-dependent protein kinase 33
Chromosome Location
Chr.1: 18781764 - 18784582 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333450.1 NP_001320603.1 6 - 7 1916 117
NM_103952.4 NP_175485.1 7 - 8 1933 117
GenBank mRNA AK175623.1 - 7 - 8 1783 117
BT012659.1 - 7 - 8 1483 117

View Full Product Details