Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KNAT4
Entrez Gene ID
830973
Gene Name
homeobox protein knotted-1-like 4
Gene Aliases
AT5G11060, KNOTTED1-like homeobox gene 4, T5K6.50, T5K6_50
Chromosome Location
Chr.5: 3509833 - 3513741 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121144.4 NP_196667.2 2 - 3 1267 73
GenBank mRNA AK118296.1 - 2 - 3 982 73

View Full Product Details