The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_121205.4 | NP_196728.1 | 16 - 17 | 1823 | 64 |
GenBank mRNA | AF428407.1 | - | 16 - 17 | 1561 | 64 |