Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MED19A
- Entrez Gene ID
-
831097
- Gene Name
- mediator of RNA polymerase II transcription subunit 19a-like protein
- Gene Aliases
-
AT5G12230, MXC9.19, MXC9_19
- Chromosome Location
-
Chr.5: 3953140 - 3956185 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_121261.3 |
NP_196784.1 |
6 - 7 |
1097 |
90 |
View Full Product Details