Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FIS1B
Entrez Gene ID
831114
Gene Name
Tetratricopeptide repeat (TPR)-like superfamily protein
Gene Aliases
AT5G12390, FISSION 1B
Chromosome Location
Chr.5: 4010443 - 4012134 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121277.4 NP_568272.1 3 - 4 548 119
GenBank mRNA AB195718.1 - 3 - 4 364 119
AK118241.1 - 3 - 4 453 119
BT005496.1 - 3 - 4 364 119

View Full Product Details