Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CESA10
Entrez Gene ID
817092
Gene Name
cellulose synthase A catalytic subunit 3 UDP-forming-like protein
Gene Aliases
AT2G25540, F13B15.20, F13B15_20, cellulose synthase 10
Chromosome Location
Chr.2: 10866822 - 10872243 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336002.1 NP_001318288.1 3 - 4 600 123
GenBank mRNA KJ138898.1 - 3 - 4 434 123

View Full Product Details