Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GP ALPHA 1
Entrez Gene ID
817170
Gene Name
G protein alpha subunit 1
Gene Aliases
ARABIDOPSIS THALIANA G PROTEIN ALPHA SUBUNIT 1, AT2G26300, ATGPA1, G PROTEIN ALPHA SUBUNIT, G PROTEIN ALPHA SUBUNIT 1, G protein alpha subunit 1, GPA1, T1D16.6, T1D16_6
Chromosome Location
Chr.2: 11196996 - 11201102 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336055.1 NP_001325056.1 6 - 7 1298 135
NM_128187.3 NP_180198.1 6 - 7 946 135
GenBank mRNA AF385704.1 - 6 - 7 644 135
AY093966.1 - 5 - 6 344 135
M32887.1 - 6 - 7 943 135

View Full Product Details