Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ER
Entrez Gene ID
817173
Gene Name
Leucine-rich receptor-like protein kinase family protein
Gene Aliases
AT2G26330, ERECTA, QRP1, QUANTITATIVE RESISTANCE TO PLECTOSPHAERELLA 1, T1D16.3, T1D16_3
Chromosome Location
Chr.2: 11208080 - 11214662 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128190.3 NP_180201.1 23 - 24 2498 105
GenBank mRNA AY035110.1 - 23 - 24 1807 105
FJ708701.1 - 23 - 24 1731 105
U47029.1 - 23 - 24 1781 105

View Full Product Details