Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AT2G28390
Entrez Gene ID
817387
Gene Name
SAND family protein
Gene Aliases
AT2G28390, MON1, MONENSIN SENSITIVITY1, T1B3.9, T1B3_9
Chromosome Location
Chr.2: 12139578 - 12143665 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128399.4 NP_029426.1 12 - 13 1952 95
GenBank mRNA AK118037.1 - 12 - 13 1739 95
AK175575.1 - 12 - 13 1739 95
AK175699.1 - 12 - 13 1739 95
AK176064.1 - 12 - 13 1749 95
AK176154.1 - 12 - 13 1749 95
AK221211.1 - 12 - 13 1739 95
AY056801.1 - 12 - 13 1732 95
AY133524.1 - 12 - 13 1662 95

View Full Product Details