Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- EXO70H5
- Entrez Gene ID
-
817413
- Gene Name
- exocyst subunit exo70 family protein H5
- Gene Aliases
-
AT2G28640, ATEXO70H5, T8O18.7, T8O18_7, exocyst subunit exo70 family protein H5
- Chromosome Location
-
Chr.2: 12284281 - 12286842 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_128425.3 |
NP_180432.2 |
1 - 2 |
1256 |
75 |
View Full Product Details