Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CERK1
- Entrez Gene ID
-
821717
- Gene Name
- chitin elicitor receptor kinase 1
- Gene Aliases
-
AT3G21630, AtCERK1, AtLYK1, LYK1, LYSM DOMAIN RECEPTOR-LIKE KINASE 1, LYSM RLK1, LysM-containing receptor-like kinase 1, chitin elicitor receptor kinase 1
- Chromosome Location
-
Chr.3: 7615357 - 7618671 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_113058.4 |
NP_566689.2 |
9 - 10 |
1594 |
125 |
GenBank mRNA |
AB367524.1 |
- |
9 - 10 |
1453 |
125 |
View Full Product Details