Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APS1
Entrez Gene ID
821861
Gene Name
ATP sulfurylase 1
Gene Aliases
AT3G22890, ATP sulfurylase 1
Chromosome Location
Chr.3: 8112614 - 8115024 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113189.5 NP_188929.1 1 - 2 724 78
GenBank mRNA AF370492.1 - 1 - 2 615 78
AK226559.1 - 1 - 2 573 78
AY050869.1 - 1 - 2 561 78
AY054169.1 - 1 - 2 573 78
AY064648.1 - 1 - 2 501 78
AY091207.1 - 1 - 2 501 78
BT002538.1 - 1 - 2 572 78

View Full Product Details