Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ETR2
- Entrez Gene ID
-
821891
- Gene Name
- Signal transduction histidine kinase, hybrid-type, ethylene sensor
- Gene Aliases
-
AT3G23150, ethylene response 2
- Chromosome Location
-
Chr.3: 8254559 - 8257888 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_113216.3 |
NP_188956.1 |
2 - 3 |
2268 |
86 |
View Full Product Details