Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TOPII
Entrez Gene ID
821972
Gene Name
topoisomerase II
Gene Aliases
AT3G23890, ATTOPII, TOPOISOMERASE II, topoisomerase II
Chromosome Location
Chr.3: 8624636 - 8631392 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125214.2 NP_001118686.1 15 - 16 3795 69
NM_113294.3 NP_189031.1 15 - 16 3805 69
GenBank mRNA L21015.1 - 15 - 16 3691 69

View Full Product Details